This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TFEC SIRNA Assays ›
Gene Symbol:
Tfec
Gene Name:
transcription factor EC
Gene Aliases:
BB107417, Tcfec, bHLHe34
Chromosome Location:
Chr.6: 16830837 - 16898462 on Build GRCm38
UniGene ID:
Mm.36217
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s74864

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tfec
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031198.3 NP_112475.1 2, 3 438
GenBank mRNA AF077742.1 - 2, 3 438
AK137141.1 - 2, 3 403
AK156327.1 - 2, 3 351
AK157426.1 - 2, 3 438
AK159579.1 - 2, 3 371
BC052832.1 - 2, 3 402
BC064770.1 - 2, 3 390
BC098494.1 - 2, 3 323 6431607

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
cellular response to heat
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
protein dimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-57b588f4f5-bcf2z:80/100.66.76.251:80.
git-commit: 377debcfccf3d85e21e16e6539a0b18561fa37ae
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.0-Offline