This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TLX1 SIRNA Assays ›
Gene Symbol:
Tlx1
Gene Name:
T cell leukemia, homeobox 1
Gene Aliases:
Hox-11, Hox11
Chromosome Location:
Chr.19: 45150079 - 45156943 on Build GRCm38
UniGene ID:
Mm.391203
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s75218

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 404.00
Your Price
Online offer: 391.65
391.65
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tlx1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021901.3 NP_068701.1 1 746
GenBank mRNA BC018246.1 - 1 746 4164519

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_001195517, NM_005521, S38742, M62626, M75952, and others.
This siRNA also targets the following transcripts in Rat: NM_001109166, XM_006231554.

Gene Ontology Categories:

Function(s) Process(es)

transcription from RNA polymerase II promoter
central nervous system development
positive regulation of cell proliferation
neuron differentiation
cell fate commitment
positive regulation of transcription from RNA polymerase II promoter
lymph node development
spleen development
organ formation
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
protein heterodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline