siRNA Details
| Gene Symbol: Tnnt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277903.1 | NP_001264832.1 | 8 | 304 | ||
| NM_001277904.1 | NP_001264833.1 | 7 | 268 | |||
| NM_011618.2 | NP_035748.1 | 8 | 301 | |||
| GenBank mRNA | AF020946.1 | - | 8 | 288 | ||
| AJ131711.1 | - | 7 | 249 | |||
| AK160226.1 | - | 8 | 302 | |||
| BC138871.1 | - | 8 | 281 | 8861893 | ||
| BC141142.1 | - | 8 | 284 | 9055754 | ||
| BC145449.1 | - | 7 | 248 | 9054007 | ||
| BC145450.1 | - | 7 | 248 | 9054008 | ||
| BC145451.1 | - | 8 | 284 | 9054009 | ||
| U92883.1 | - | 8 | 285 | |||
| U92884.1 | - | 7 | 252 | |||

Tnnt1
Genomic Map