siRNA Details
| Gene Symbol: Bace1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145947.1 | NP_001139419.1 | 8 | 1766 | ||
| NM_011792.5 | NP_035922.4 | 9 | 1868 | |||
| GenBank mRNA | AF190726.2 | - | 9 | 1796 | ||
| AF200346.1 | - | 9 | 1369 | |||
| AK014390.2 | - | 5 | 667 | |||
| AK014464.1 | - | 9 | 1798 | |||
| AK033112.1 | - | 9 | 1818 | |||
| AK041285.1 | - | 9 | 1818 | |||
| AK046175.1 | - | 9 | 1815 | |||
| AK049626.1 | - | 9 | 1801 | |||
| AK079814.1 | - | 8 | 1086 | |||
| AK080498.1 | - | 9 | 1820 | |||
| AK082230.1 | - | 9 | 1818 | |||
| AK082317.1 | - | 8 | 1717 | |||
| AK163583.1 | - | 9 | 1819 | |||
| AK173112.1 | - | 7 | 986 | |||
| BC048189.1 | - | 9 | 1794 | 6831622 | ||

Bace1
Genomic Map