siRNA Details
| Gene Symbol: Slc26a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011867.3 | NP_035997.1 | 6 | 914 | ||
| GenBank mRNA | AF167411.1 | - | 6 | 915 | ||
| BC140995.1 | - | 5 | 762 | 9055607 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc26a4| Gene Symbol: Slc26a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011867.3 | NP_035997.1 | 6 | 914 | ||
| GenBank mRNA | AF167411.1 | - | 6 | 915 | ||
| BC140995.1 | - | 5 | 762 | 9055607 | ||