This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TLK2 SIRNA Assays ›
Gene Symbol:
Tlk2
Gene Name:
tousled-like kinase 2 (Arabidopsis)
Gene Aliases:
4933403M19Rik, PKU-alpha, PKUalpha, Tlk
Chromosome Location:
Chr.11: 105167521 - 105283959 on Build GRCm38
UniGene ID:
Mm.126976
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s76894

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 400.00
Your Price
Online offer: 296.00
296.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tlk2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001112705.2 NP_001106176.1 18 2169
NM_001294329.1 NP_001281258.1 16 2029
NM_001294330.1 NP_001281259.1 16 1977
NM_001294331.1 NP_001281260.1 18 2182
NM_001294334.1 NP_001281263.1 18 2169
GenBank mRNA AF045252.1 - 18 2188
AF045253.1 - 17 1616
AF045254.1 - 13 1330
AK014829.1 - 18 1807
AK089925.1 - 16 1557
AK165667.1 - 18 2172
AK211763.1 - 2 163
AX775743.1 - 12 1194
BC066198.1 - 16 1522
U56909.1 - 9 867

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001191652, XM_006247576, XM_008768350, XM_006247574, XM_008768348, and others.

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

regulation of chromatin assembly or disassembly
protein phosphorylation
cellular response to DNA damage stimulus
cell cycle
chromosome segregation
multicellular organism development
spermatogenesis
phosphorylation
chromatin modification
peptidyl-serine phosphorylation
cell differentiation
negative regulation of proteasomal ubiquitin-dependent protein catabolic process
intracellular signal transduction
cellular response to gamma radiation
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-nlc25:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline