This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other MAP2K3 SIRNA Assays ›
Gene Symbol:
Map2k3
Gene Name:
mitogen-activated protein kinase kinase 3
Gene Aliases:
AW212142, MEK3, MKK3, Prkmk3, mMKK3b
Chromosome Location:
Chr.11: 60932057 - 60952803 on Build GRCm38
UniGene ID:
Mm.18494
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s77059

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Map2k3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008928.4 NP_032954.1 9 903
GenBank mRNA AK008141.1 - 9 906
AK011002.1 - 9 904
AK053378.1 - 9 904
AK150067.1 - 9 904
AK159379.1 - 9 902
AK166255.1 - 9 905
AK166507.1 - 9 904
AK167209.1 - 9 904
AK167442.1 - 9 907
AK168710.1 - 9 886
BC007467.1 - 8 842 3497714
D87115.1 - 9 916
X93150.1 - 7 638

Back To Top

More Information


Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

MAPK cascade
activation of MAPK activity
protein phosphorylation
inflammatory response
regulation of mitotic cell cycle
phosphorylation
stress-activated protein kinase signaling cascade
regulation of cytokine biosynthetic process
regulation of apoptotic process
positive regulation of protein kinase activity
positive regulation of transcription, DNA-templated
cardiac muscle contraction
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
MAP kinase kinase activity
protein tyrosine kinase activity
protein binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
protein kinase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mjsjv:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline