siRNA Details
| Gene Symbol: St6galnac5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012028.4 | NP_036158.3 | 5 | 993 | ||
| GenBank mRNA | AB028840.1 | - | 975 | |||
| AB030836.1 | - | 5 | 996 | |||
| AK034387.1 | - | 997 | ||||
| AK038434.1 | - | 5 | 997 | |||
| AK042683.1 | - | 5 | 1033 | |||
| BC055737.1 | - | 5 | 993 | 6415191 | ||

St6galnac5
Genomic Map