This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other FBXL17 SIRNA Assays ›
Gene Symbol:
Fbxl17
Gene Name:
F-box and leucine-rich repeat protein 17
Gene Aliases:
6330576B01Rik, AI452053, BB073797, C130023C01Rik, Fbx13, Fbxo13
Chromosome Location:
Chr.17: 63045949 - 63504042 on Build GRCm38
UniGene ID:
Mm.136366
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s78415

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fbxl17
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015794.1 NP_056609.1 8 2441
GenBank mRNA AF176529.1 - 8 1161
AK004419.1 - 2 123
AK154497.1 - 8 950
BC058120.1 - 5 499 6810624
BC138330.1 - 5 477 8861350
BC138331.1 - 5 477 8861351

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-x666z:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline