siRNA Details
| Gene Symbol: Crim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015800.3 | NP_056615.1 | 7, 8 | 1901 | ||
| GenBank mRNA | AF168680.1 | - | 7, 8 | 1353 | ||
| AK053197.1 | - | 5, 6 | 844 | |||
| BC100348.1 | - | 7, 8 | 1686 | 30521148 | ||

Crim1
Genomic Map