This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other DDAH2 SIRNA Assays ›
Gene Symbol:
Ddah2
Gene Name:
dimethylarginine dimethylaminohydrolase 2
Gene Aliases:
1110003M04Rik, AU019324, AW413173, DDAH-2, DDAHII, Ddah, G6a, NG30
Chromosome Location:
Chr.17: 35059035 - 35062099 on Build GRCm38
UniGene ID:
Mm.1457
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s78652

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ddah2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001190449.1 NP_001177378.1 5 787
NM_016765.3 NP_058045.1 5 787
GenBank mRNA AF004106.1 - 5 721
BC003328.1 - 5 737 3158175

Back To Top

More Information


Panther Classification:

Molecular Function -

hydrolase

Gene Ontology Categories:

Function(s) Process(es)

citrulline metabolic process
arginine metabolic process
arginine catabolic process
dimethylargininase activity
amino acid binding
hydrolase activity
hydrolase activity, acting on carbon-nitrogen (but not peptide) bonds, in linear amidines

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0