This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PSMD10 SIRNA Assays ›
Gene Symbol:
Psmd10
Gene Name:
proteasome (prosome, macropain) 26S subunit, non-ATPase, 10
Gene Aliases:
AW554874
Chromosome Location:
Chr.X: 140948425 - 140956711 on Build GRCm38
UniGene ID:
Mm.17640
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s79154

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Psmd10
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164177.1 NP_001157649.1 3, 4 409
NM_016883.4 NP_058579.2 3, 4 409
GenBank mRNA AK009068.1 - 3, 4 388
AK018233.1 - 3, 4 387
AK136400.1 - 3, 4 388
BC026931.1 - 3, 4 376 3987112
BC056196.1 - 3, 4 394 30019985
CD564300.1 - 3, 4 376

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_053925, AB022014.

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
apoptotic process
cytoplasmic sequestering of NF-kappaB
positive regulation of cell growth
positive regulation of protein ubiquitination
negative regulation of NF-kappaB transcription factor activity
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
negative regulation of apoptotic process
negative regulation of MAPK cascade
negative regulation of DNA damage response, signal transduction by p53 class mediator
positive regulation of cyclin-dependent protein serine/threonine kinase activity
proteasome regulatory particle assembly
negative regulation of release of cytochrome c from mitochondria
transcription factor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7669c5b596-vc5lg:80/100.66.78.61:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0