This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other BYSL SIRNA Assays ›
Gene Symbol:
Bysl
Gene Name:
bystin-like
Gene Aliases:
Bys, Enp1
Chromosome Location:
Chr.17: 47599331 - 47611492 on Build GRCm38
UniGene ID:
Mm.472101
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s79167

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 400.00
Your Price
Online offer: 296.00
296.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Bysl
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016859.3 NP_058555.3 3, 4 889
GenBank mRNA AF007802.1 - 1, 2 118
AK019239.1 - 3, 4 624
AK032993.1 - 3, 4 621
AK049195.1 - 3, 4 882
AK051704.1 - 3, 4 879
AK083033.1 - 3, 4 876
AK143027.1 - 3, 4 890
AK168301.1 - 3, 4 627
BC017530.1 - 3, 4 616 4925307

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_182674, AY257675, CV104390, BC079030.

Gene Ontology Categories:

Function(s) Process(es)

maturation of SSU-rRNA from tricistronic rRNA transcript (SSU-rRNA, 5.8S rRNA, LSU-rRNA)
in utero embryonic development
blastocyst formation
trophectodermal cell differentiation
cell adhesion
cell proliferation
ribosome biogenesis
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-hssmq:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline