siRNA Details
| Gene Symbol: Tmem131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018872.2 | NP_061360.2 | 20 | 2326 | ||
| GenBank mRNA | AB093221.1 | - | 16 | 1762 | ||
| AF060565.1 | - | 20 | 2028 | |||
| BU058584.1 | - | 4 | 431 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem131| Gene Symbol: Tmem131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018872.2 | NP_061360.2 | 20 | 2326 | ||
| GenBank mRNA | AB093221.1 | - | 16 | 1762 | ||
| AF060565.1 | - | 20 | 2028 | |||
| BU058584.1 | - | 4 | 431 | |||