siRNA Details
| Gene Symbol: Astn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019514.3 | NP_062387.2 | 14 | 2593 | ||
| NM_207109.2 | NP_996992.1 | 15 | 2749 | |||
| GenBank mRNA | AF116573.1 | - | 12 | 1761 | ||
| AK041160.1 | - | 16 | 2653 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Astn2| Gene Symbol: Astn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019514.3 | NP_062387.2 | 14 | 2593 | ||
| NM_207109.2 | NP_996992.1 | 15 | 2749 | |||
| GenBank mRNA | AF116573.1 | - | 12 | 1761 | ||
| AK041160.1 | - | 16 | 2653 | |||