This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TDP2 SIRNA Assays ›
Gene Symbol:
Tdp2
Gene Name:
tyrosyl-DNA phosphodiesterase 2
Gene Aliases:
D13Ertd656e, Ttrap
Chromosome Location:
Chr.13: 24831647 - 24842153 on Build GRCm38
UniGene ID:
Mm.427162
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s80151

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 400.00
Your Price
Online offer: 388.65
388.65
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tdp2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019551.2 NP_062424.1 6 817
GenBank mRNA AJ251328.1 - 6 791
AK009089.1 - 6 791
AK135925.1 - 6 829
AK167707.1 - 5 613
BC132511.1 - 6 780 40130788
BC132513.1 - 6 780 40130790
BC145334.1 - 5 609

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

DNA repair
double-strand break repair
cellular response to DNA damage stimulus
nucleic acid phosphodiester bond hydrolysis
magnesium ion binding
single-stranded DNA binding
cyclic-nucleotide phosphodiesterase activity
3',5'-cyclic-AMP phosphodiesterase activity
calmodulin-dependent cyclic-nucleotide phosphodiesterase activity
cGMP-stimulated cyclic-nucleotide phosphodiesterase activity
cGMP-inhibited cyclic-nucleotide phosphodiesterase activity
photoreceptor cyclic-nucleotide phosphodiesterase activity
nuclease activity
endonuclease activity
phosphoric diester hydrolase activity
hydrolase activity
manganese ion binding
tyrosyl-RNA phosphodiesterase activity
7,8-dihydro-D-neopterin 2',3'-cyclic phosphate phosphodiesterase activity
metal ion binding
inositol phosphosphingolipid phospholipase activity
inositol phosphorylceramide phospholipase activity
mannosyl-inositol phosphorylceramide phospholipase activity
mannosyl-diinositol phosphorylceramide phospholipase activity
5'-tyrosyl-DNA phosphodiesterase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-875b6bdcc-9td46:80/100.66.75.104:80.
git-commit: 41fbaa8c9960493fb804edf63312caa25f4bdf16
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0