This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PDCD10 SIRNA Assays ›
Gene Symbol:
Pdcd10
Gene Name:
programmed cell death 10
Gene Aliases:
2410003B13Rik, Ccm3, Tfa15, Tfar15
Chromosome Location:
Chr.3: 75516490 - 75556852 on Build GRCm38
UniGene ID:
Mm.316473
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s80539

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pdcd10
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019745.4 NP_062719.2 2, 3 357
GenBank mRNA AF159368.1 - 2, 3 260
AK005594.1 - 2, 3 286
AK075926.1 - 2, 3 305
AK077308.1 - 2, 3 334
AK137198.1 - 2, 3 327
AK149182.1 - 2, 3 263
AK160234.1 - 2, 3 307
AK168857.1 - 2, 3 291
AK169689.1 - 2, 3 291
BC019650.1 - 2, 3 297 4022513
BC086778.1 - 2, 3 276 5689088

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
apoptotic process
positive regulation of cell proliferation
positive regulation of gene expression
negative regulation of gene expression
positive regulation of cell migration
positive regulation of stress-activated MAPK cascade
positive regulation of peptidyl-serine phosphorylation
intrinsic apoptotic signaling pathway in response to hydrogen peroxide
response to hydrogen peroxide
negative regulation of apoptotic process
positive regulation of MAP kinase activity
wound healing, spreading of cells
positive regulation of Notch signaling pathway
protein stabilization
establishment of Golgi localization
positive regulation of protein serine/threonine kinase activity
negative regulation of cell migration involved in sprouting angiogenesis
Golgi reassembly
negative regulation of blood vessel endothelial cell proliferation involved in sprouting angiogenesis
protein binding
protein kinase binding
protein homodimerization activity
protein N-terminus binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-f8k5r:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline