This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other MBNL1 SIRNA Assays ›
Gene Symbol:
Mbnl1
Gene Name:
muscleblind-like 1 (Drosophila)
Gene Aliases:
Mbnl, mKIAA0428
Chromosome Location:
Chr.3: 60472791 - 60629750 on Build GRCm38
UniGene ID:
Mm.255723
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s80972

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mbnl1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001253708.1 NP_001240637.1 2 1103
NM_020007.3 NP_064391.2 2 1103
GenBank mRNA AB093240.1 - 2 1107
AF231110.1 - 1 543
AK038242.1 - 1 138
AK088871.1 - 2 1094
BC060031.1 - 2 1081

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001191566, XM_008761040, XM_008761037, XM_008761042, XM_008761032, and others.

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

alternative mRNA splicing, via spliceosome
regulation of alternative mRNA splicing, via spliceosome
mRNA splice site selection
mRNA processing
skeletal muscle tissue development
RNA splicing
regulation of RNA splicing
myoblast differentiation
RNA binding
double-stranded RNA binding
poly(A) RNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mjsjv:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline