siRNA Details
| Gene Symbol: Park7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020569.3 | NP_065594.2 | 4 | 386 | ||
| GenBank mRNA | AB015652.1 | - | 4 | 333 | ||
| AK146368.1 | - | 4 | 298 | |||
| AK153948.1 | - | 4 | 343 | |||
| AK168341.1 | - | 4 | 316 | |||
| BC002187.1 | - | 4 | 292 | 3487763 | ||
| CA456587.1 | - | 4 | 424 | |||

Park7
Genomic Map