siRNA Details
| Gene Symbol: Park7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020569.3 | NP_065594.2 | 3, 4 | 345 | ||
| GenBank mRNA | AB015652.1 | - | 3, 4 | 292 | ||
| AK146368.1 | - | 3, 4 | 257 | |||
| AK153948.1 | - | 3, 4 | 302 | |||
| AK168341.1 | - | 3, 4 | 275 | |||
| BC002187.1 | - | 3, 4 | 251 | 3487763 | ||
| CA456587.1 | - | 3, 4 | 383 | |||

Park7
Genomic Map