siRNA Details
| Gene Symbol: Noc2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021303.2 | NP_067278.2 | 17 | 1984 | ||
| GenBank mRNA | AF155546.1 | - | 5 | 494 | ||
| AK076287.1 | - | 16 | 2026 | |||
| AK078560.1 | - | 17 | 1962 | |||
| AK133834.1 | - | 17 | 1984 | |||
| AK157765.1 | - | 17 | 1977 | |||
| BC020013.1 | - | 17 | 1955 | 3663482 | ||

Noc2l
Genomic Map