This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other HIC2 SIRNA Assays ›
Gene Symbol:
Hic2
Gene Name:
hypermethylated in cancer 2
Gene Aliases:
AA409108, HRG22, mKIAA1020
Chromosome Location:
Chr.16: 17233587 - 17263430 on Build GRCm38
UniGene ID:
Mm.261602
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s81509

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Hic2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178922.3 NP_849253.2 3 755
GenBank mRNA AJ868292.1 - 2 719
AK028357.1 - 2 567
AK084975.1 - 3 678
AK129266.1 - 3 741
AK156332.1 - 3 771
AK157276.1 - 3 771
BC065124.1 - 754 30537019

Back To Top

More Information


Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
nucleic acid binding
DNA binding
protein C-terminus binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-cj8gb:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline