This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other RNF130 SIRNA Assays ›
Gene Symbol:
Rnf130
Gene Name:
ring finger protein 130
Gene Aliases:
2510042A13Rik, G1RZFP, G1rp, GOLIATH, GP
Chromosome Location:
Chr.11: 50025331 - 50125719 on Build GRCm38
UniGene ID:
Mm.359004
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s81854

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 400.00
Your Price
Online offer: 296.00
296.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rnf130
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290749.1 NP_001277678.1 4 824
NM_001290750.1 NP_001277679.1 4 824
NM_021540.4 NP_067515.2 4 824
GenBank mRNA AF171875.1 - 4 792
AK011088.1 - 4 809
AK220529.1 - 4 794
BC018199.1 - 4 818 4017435
BC048901.1 - 4 807 6390079

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001037658, AY190520, BC166595, FQ234228.

Gene Ontology Categories:

Function(s) Process(es)

apoptotic process
programmed cell death
protein ubiquitination
ubiquitin-protein transferase activity
zinc ion binding
UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
ligase activity
ribosomal S6-glutamic acid ligase activity
coenzyme F420-0 gamma-glutamyl ligase activity
coenzyme F420-2 alpha-glutamyl ligase activity
metal ion binding
protein-glycine ligase activity
protein-glycine ligase activity, initiating
protein-glycine ligase activity, elongating
tubulin-glycine ligase activity
protein-glutamic acid ligase activity
tubulin-glutamic acid ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-7wnc4:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline