siRNA Details
| Gene Symbol: Stk32b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022416.2 | NP_071861.1 | 1, 2 | 417 | ||
| GenBank mRNA | AJ250840.1 | - | 1, 2 | 495 | ||
| AK082468.1 | - | 1, 2 | 418 | |||
| BC052404.1 | - | 1, 2 | 327 | 5715729 | ||
| BC056396.1 | - | 1, 2 | 403 | 6841272 | ||
| BC058412.1 | - | 1, 2 | 327 | 5715729 | ||

Stk32b
Genomic Map