This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TMUB1 SIRNA Assays ›
Gene Symbol:
Tmub1
Gene Name:
transmembrane and ubiquitin-like domain containing 1
Gene Aliases:
2010004O20Rik, AB030183, Hops
Chromosome Location:
Chr.5: 24445463 - 24448331 on Build GRCm38
UniGene ID:
Mm.150098
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s82188

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 404.00
Your Price
Online offer: 391.65
391.65
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tmub1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022418.3 NP_071863.1 3 545
GenBank mRNA AB030183.1 - 3 507
AK008109.1 - 3 479
AY603379.1 - 3 476
BC019547.1 - 3 501 4164765

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-55658555d6-9rjdb:80/100.66.79.224:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline