siRNA Details
| Gene Symbol: Cldn12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193659.1 | NP_001180588.1 | 4 | 902 | ||
| NM_001193660.1 | NP_001180589.1 | 5 | 850 | |||
| NM_001193661.1 | NP_001180590.1 | 4 | 737 | |||
| NM_022890.2 | NP_075028.1 | 4 | 892 | |||
| GenBank mRNA | AF247664.1 | - | 1 | 484 | ||
| AK039672.1 | - | 4 | 784 | |||
| AK049331.1 | - | 4 | 759 | |||
| AK128999.1 | - | 4 | 771 | |||
| AK132155.1 | - | 4 | 761 | |||
| AK166110.1 | - | 4 | 713 | |||
| BC024057.1 | - | 4 | 801 | 5100104 | ||

Cldn12
Genomic Map