siRNA Details
| Gene Symbol: Tpd52l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291197.1 | NP_001278126.1 | 6, 7 | 686 | ||
| NM_001291201.1 | NP_001278130.1 | 5, 6 | 626 | |||
| NM_001291202.1 | NP_001278131.1 | 5, 6 | 623 | |||
| NM_025482.3 | NP_079758.2 | 6, 7 | 683 | |||
| GenBank mRNA | AK013085.1 | - | 6, 7 | 625 | ||
| AK013188.1 | - | 6, 7 | 636 | |||
| AK051268.1 | - | 5, 6 | 567 | |||
| AK159925.1 | - | 6, 7 | 611 | |||
| BC006886.1 | - | 6, 7 | 572 | 2647702 | ||
| DV047279.1 | - | 6, 7 | 672 | |||

Tpd52l2
Genomic Map