This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other AQP11 SIRNA Assays ›
Gene Symbol:
Aqp11
Gene Name:
aquaporin 11
Gene Aliases:
1700015P13Rik, AI648923, sjds
Chromosome Location:
Chr.7: 97725490 - 97738514 on Build GRCm38
UniGene ID:
Mm.29756
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s83038

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 400.00
Your Price
Online offer: 296.00
296.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Aqp11
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175105.3 NP_780314.1 1 687
GenBank mRNA AB028148.1 - 1 441
AK006005.1 - 1 712
AK040045.1 - 1 456
AK075757.1 - 1 688
AK137326.1 - 1 731
BC037088.1 - 1 466
BC068103.1 - 2 336

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

kidney development
transport
water transport
endosomal lumen acidification
protein homooligomerization
transmembrane transport
proximal tubule development
transporter activity
water channel activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mlg7h:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline