siRNA Details
| Gene Symbol: Aptx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025444.3 | NP_001020615.1 | 2 | 169 | ||
| NM_001025445.2 | NP_001020616.1 | 2 | 169 | |||
| NM_025545.4 | NP_079821.3 | 2 | 304 | |||
| GenBank mRNA | AK005286.1 | - | 1 | 121 | ||
| AK010516.1 | - | 2 | 156 | |||
| AK017522.1 | - | 1 | 139 | |||
| AK077351.1 | - | 2 | 305 | |||
| AK088928.1 | - | 2 | 139 | |||
| AY040780.1 | - | 2 | 315 | |||
| AY208844.1 | - | 2 | 316 | |||
| BB860888.1 | - | 2 | 173 | |||
| BC021872.1 | - | 2 | 162 | 3994375 | ||
| BC068309.1 | - | 1 | 82 | |||
| BC071213.1 | - | 2 | 156 | |||

Aptx
Genomic Map