This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other 2410004B18RIK SIRNA Assays ›
Gene Symbol:
2410004B18Rik
Gene Name:
RIKEN cDNA 2410004B18 gene
Gene Aliases:
4930572C24Rik
Chromosome Location:
Chr.3: 145938003 - 145944829 on Build GRCm38
UniGene ID:
Mm.25137
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s83233

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 2410004B18Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025555.4 NP_079831.1 3 514
GenBank mRNA AK010378.1 - 3 514
AK019404.1 - 3 510
AK019806.1 - 2 158
AK077565.1 - 4 632
BC049577.1 - 4 468
BC115465.2 - 3 513 40100373

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline