This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SLC30A7 SIRNA Assays ›
Gene Symbol:
Slc30a7
Gene Name:
solute carrier family 30 (zinc transporter), member 7
Gene Aliases:
1810059J10Rik, 2610034N15Rik, 4833428C12Rik, AI467242, ZnT-7, ZnT7, Zntl2
Chromosome Location:
Chr.3: 115938973 - 116007414 on Build GRCm38
UniGene ID:
Mm.34550
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s83378

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slc30a7
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023214.7 NP_075703.1 8 934
GenBank mRNA AF233322.1 - 8 796
AF529196.1 - 8 742
AK010008.1 - 8 934
AK075802.1 - 8 907
BC017136.1 - 8 884 4911068
BC050193.1 - 8 833 30019802

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
cation transport
zinc II ion transport
transmembrane transport
zinc ion binding
cation transmembrane transporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-j47h5:80/100.66.72.248:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline