siRNA Details
| Gene Symbol: Spcs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025668.3 | NP_079944.1 | 1, 2 | 131 | ||
| GenBank mRNA | AK014310.1 | - | 1, 2 | 145 | ||
| AK017504.1 | - | 1, 2 | 136 | |||
| AK077510.1 | - | 1, 2 | 141 | |||
| AK134578.1 | - | 1, 2 | 132 | |||
| AK168173.1 | - | 1, 2 | 136 | |||
| BC050150.1 | - | 1, 2 | 141 | 30026746 | ||

Spcs2
Genomic Map