This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ALG14 SIRNA Assays ›
Gene Symbol:
Alg14
Gene Name:
asparagine-linked glycosylation 14
Gene Aliases:
5430428G01Rik, AI854024
Chromosome Location:
Chr.3: 121291716 - 121362980 on Build GRCm38
UniGene ID:
Mm.269881
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s83858

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 400.00
Your Price
Online offer: 296.00
296.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Alg14
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024178.2 NP_077140.1 2 312
GenBank mRNA AK011733.1 - 2 358
AK017354.1 - 1 13
BC002278.1 - 2 312 3496897

Back To Top

More Information


Panther Classification:

Molecular Function -

transferase

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-9hn7z:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline