This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CNTNAP2 SIRNA Assays ›
Gene Symbol:
Cntnap2
Gene Name:
contactin associated protein-like 2
Gene Aliases:
5430425M22Rik, Caspr2, mKIAA0868
Chromosome Location:
Chr.6: 45060061 - 47301388 on Build GRCm38
UniGene ID:
Mm.440084
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s83871

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cntnap2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004357.2 NP_001004357.2 24 4188
NM_025771.3 NP_080047.1 4 635
GenBank mRNA AK013139.1 - 4 603
AK017341.1 - 4 630
AK129232.1 - 24 4161
BC029826.1 - 4 584
BC064467.1 - 19 3079
BC103799.1 - 4 542
BC132074.1 - 3 447
BC132076.1 - 3 447

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_014141, AB020675, BC093780, AF319045, AB073883, and others.
This siRNA also targets the following transcripts in Rat: XM_008762930.

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
learning
neuron recognition
adult behavior
neuron projection development
social behavior
vocal learning
clustering of voltage-gated potassium channels
protein localization to juxtaparanode region of axon
vocalization behavior
protease binding
protein binding
enzyme binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gjvvj:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0