siRNA Details
| Gene Symbol: Unc50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026123.3 | NP_080399.1 | 2 | 367 | ||
| GenBank mRNA | AK003273.1 | - | 2 | 142 | ||
| AK004433.1 | - | 2 | 131 | |||
| AK132757.1 | - | 2 | 368 | |||
| AK135885.1 | - | 2 | 359 | |||
| AK217920.1 | - | 2 | 88 | |||
| AY017214.1 | - | 2 | 352 | |||
| BC019484.1 | - | 2 | 334 | 4039903 | ||

Unc50
Genomic Map