This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other 4930549C01RIK SIRNA Assays ›
Gene Symbol:
4930549C01Rik
Gene Name:
RIKEN cDNA 4930549C01 gene
Gene Aliases:
-
Chromosome Location:
Chr.4: 136606944 - 136613277 on Build GRCm38
UniGene ID:
Mm.158168
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s85420

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 4930549C01Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026300.2 NP_080576.1 2 417
GenBank mRNA AK015053.1 - 2 380
AK016074.1 - 2 418
BC048442.1 - 2 407 6704947

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-875b6bdcc-hzcnf:80/100.66.78.54:80.
git-commit: 41fbaa8c9960493fb804edf63312caa25f4bdf16
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0