This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SDHB SIRNA Assays ›
Gene Symbol:
Sdhb
Gene Name:
succinate dehydrogenase complex, subunit B, iron sulfur (Ip)
Gene Aliases:
0710008N11Rik
Chromosome Location:
Chr.4: 140961271 - 140979197 on Build GRCm38
UniGene ID:
Mm.246965
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s85449

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Sdhb
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023374.3 NP_075863.2 2, 3 337
GenBank mRNA AK003052.1 - 2, 3 231
AK003533.1 - 2, 3 221
AK009660.1 - 2, 3 231
AK153045.1 - 2, 3 214
AK169252.1 - 2, 3 228
AK169839.1 - 2, 3 230
BC051934.1 - 2, 3 337 5688898

Back To Top

More Information


Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

tricarboxylic acid cycle
succinate metabolic process
transport
respiratory electron transport chain
oxidation-reduction process
succinate dehydrogenase (ubiquinone) activity
electron carrier activity
oxidoreductase activity
metal ion binding
iron-sulfur cluster binding
2 iron, 2 sulfur cluster binding
3 iron, 4 sulfur cluster binding
4 iron, 4 sulfur cluster binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0