This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other 2700060E02RIK SIRNA Assays ›
Gene Symbol:
2700060E02Rik
Gene Name:
RIKEN cDNA 2700060E02 gene
Gene Aliases:
-
Chromosome Location:
Chr.14: 19811402 - 19823823 on Build GRCm38
UniGene ID:
Mm.21932
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s86145

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 400.00
Your Price
Online offer: 296.00
296.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 2700060E02Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026528.3 NP_080804.1 6 600
GenBank mRNA AK003732.1 - 6 596
AK012451.1 - 6 564
AK012460.1 - 6 595
AK012486.1 - 6 601
AK148686.1 - 6 542
AK160555.1 - 6 564
AK160696.1 - 6 602
AK162070.1 - 6 552
AK162433.1 - 6 529
BC023932.1 - 6 579
BC048357.1 - 6 595 3979120
BC096571.1 - 6 583 963861

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001047874, XM_006251636, FQ229497, FQ215371, AY539943, and others.

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
tRNA splicing, via endonucleolytic cleavage and ligation
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II core binding
RNA binding
identical protein binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mjsjv:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline