This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SYMPK SIRNA Assays ›
Gene Symbol:
Sympk
Gene Name:
symplekin
Gene Aliases:
1500016F02Rik, 4632415H16Rik, AA125406, AI449890, SPK, SYM
Chromosome Location:
Chr.7: 19024352 - 19054623 on Build GRCm38
UniGene ID:
Mm.130902
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s86360

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Sympk
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026605.2 NP_080881.2 14 2101
GenBank mRNA AK019497.1 - 9 1577
AK146894.1 - 14 2082
AK155093.1 - 14 2102
AK155375.1 - 14 2056
AK165591.1 - 14 2080
AK168541.1 - 14 2082
AK168757.1 - 14 2082
AK172040.1 - 15 2130
AK215540.1 - 2 238
BC049852.1 - 14 2046 4163760

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

mRNA polyadenylation
mRNA processing
cell adhesion
positive regulation of protein dephosphorylation
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-ftbch:80/100.66.79.106:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0