siRNA Details
| Gene Symbol: 1110012L19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026787.2 | NP_081063.1 | 3 | 624 | ||
| GenBank mRNA | AK003642.1 | - | 3 | 627 | ||
| BC024574.1 | - | 3 | 569 | 5042031 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1110012L19Rik| Gene Symbol: 1110012L19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026787.2 | NP_081063.1 | 3 | 624 | ||
| GenBank mRNA | AK003642.1 | - | 3 | 627 | ||
| BC024574.1 | - | 3 | 569 | 5042031 | ||