This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SPCS1 SIRNA Assays ›
Gene Symbol:
Spcs1
Gene Name:
signal peptidase complex subunit 1 homolog (S. cerevisiae)
Gene Aliases:
1810004F21Rik
Chromosome Location:
Chr.14: 30999826 - 31001666 on Build GRCm38
UniGene ID:
Mm.28852
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s87339

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Spcs1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026911.3 NP_081187.2 3 569
GenBank mRNA AK007336.1 - 3 267
AK082928.1 - 3 268
AK088658.1 - 2 352
BC020050.1 - 471
BC034270.2 - 3 444
BC049895.1 - 3 260 3991764

Back To Top

More Information


Panther Classification:

Molecular Function -

protease

Gene Ontology Categories:

Function(s) Process(es)

signal peptide processing
proteolysis
biological_process
protein targeting to ER
peptidase activity
cysteine-type peptidase activity
hydrolase activity
ribosome binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-7wnc4:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline