siRNA Details
| Gene Symbol: Spcs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026911.3 | NP_081187.2 | 2, 3 | 529 | ||
| GenBank mRNA | AK007336.1 | - | 2, 3 | 227 | ||
| AK082928.1 | - | 2, 3 | 228 | |||
| BC034270.2 | - | 2, 3 | 404 | |||
| BC049895.1 | - | 2, 3 | 220 | 3991764 | ||
| BX512708.1 | - | 2, 3 | 471 | |||

Spcs1
Genomic Map