siRNA Details
| Gene Symbol: SCGB2A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002411.3 | NP_002402.1 | 2 | 210 | ||
| XM_005274005.3 | XP_005274062.3 | 2 | 235 | |||
| GenBank mRNA | AI904278.1 | - | 2 | 210 | ||
| AY217100.1 | - | 2 | 148 | |||
| BC067220.1 | - | 2 | 211 | 30321681 | ||
| BC128252.1 | - | 2 | 202 | 40115103 | ||
| BC128402.1 | - | 2 | 202 | 40115099 | ||
| U33147.1 | - | 2 | 208 | |||

SCGB2A2
Genomic Map