This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SPATA4 SIRNA Assays ›
Gene Symbol:
Spata4
Gene Name:
spermatogenesis associated 4
Gene Aliases:
1700001N01Rik, Srg2, TSARG2
Chromosome Location:
Chr.8: 54600781 - 54610098 on Build GRCm38
UniGene ID:
Mm.45833
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s87650

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Spata4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133711.3 NP_598472.2 4 528
GenBank mRNA AF395083.2 - 4 528
AF530515.1 - 4 528
AK005612.1 - 4 528
BC048650.1 - 4 528 6772684

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-9hvhj:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline