siRNA Details
| Gene Symbol: Asb9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027027.2 | NP_081303.1 | 577 | |||
| GenBank mRNA | AF398970.1 | - | 4 | 509 | ||
| AK005878.1 | - | 578 | ||||
| BC061038.1 | - | 569 | 6703903 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Asb9| Gene Symbol: Asb9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027027.2 | NP_081303.1 | 577 | |||
| GenBank mRNA | AF398970.1 | - | 4 | 509 | ||
| AK005878.1 | - | 578 | ||||
| BC061038.1 | - | 569 | 6703903 | |||