This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other 1700030J22RIK SIRNA Assays ›
Gene Symbol:
1700030J22Rik
Gene Name:
RIKEN cDNA 1700030J22 gene
Gene Aliases:
-
Chromosome Location:
Chr.8: 116969594 - 116979001 on Build GRCm38
UniGene ID:
Mm.272846
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s87888

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 1700030J22Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027103.2 NP_081379.1 3 1255
GenBank mRNA AK006553.1 - 3 1260
AK030075.1 - 3 1256
AK031529.1 - 3 1140
BC049630.1 - 3 1184
BC100501.1 - 3 1275 30917058
BC137750.1 - 3 1193 8860769
BC137751.1 - 3 1193 8860770

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-5p4xw:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline