This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other KCTD2 SIRNA Assays ›
Gene Symbol:
Kctd2
Gene Name:
potassium channel tetramerisation domain containing 2
Gene Aliases:
2310012I15Rik
Chromosome Location:
Chr.11: 115420126 - 115431274 on Build GRCm38
UniGene ID:
Mm.276299
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s88770

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Kctd2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183285.3 NP_899108.3 3 559
GenBank mRNA AA537633.1 - 3 213
AK009318.1 - 3 556
BC095988.1 - 3 518 5365458

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
protein homooligomerization
protein complex binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-57b588f4f5-qj299:80/100.66.76.91:80.
git-commit: 377debcfccf3d85e21e16e6539a0b18561fa37ae
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.0-Offline