This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ECD SIRNA Assays ›
Gene Symbol:
Ecd
Gene Name:
ecdysoneless homolog (Drosophila)
Gene Aliases:
5730461K03Rik, Hsgt1
Chromosome Location:
Chr.14: 20319859 - 20348121 on Build GRCm38
UniGene ID:
Mm.35105
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s88989

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ecd
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027475.3 NP_081751.1 14 2125
GenBank mRNA AK017681.1 - 12 1549
AK028664.1 - 14 2089
AK043261.1 - 14 1974
BC013470.1 - 14 2074 4220504

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
cell proliferation
regulation of G1/S transition of mitotic cell cycle
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-2scln:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline