This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other 4931429I11RIK SIRNA Assays ›
Gene Symbol:
4931429I11Rik
Gene Name:
RIKEN cDNA 4931429I11 gene
Gene Aliases:
BB239335, jhy
Chromosome Location:
Chr.9: 40894849 - 40966864 on Build GRCm38
UniGene ID:
Mm.335087
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s89335

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 4931429I11Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081121.1 NP_001074590.1 5 1060

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

brain development
ciliary basal body organization
cerebrospinal fluid secretion
axoneme assembly
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-c565d5cb7-s92gb:80/100.66.74.196:80.
git-commit: 04d74f786bdbf98a9bc1b2a22061fbbfd0e8b2c2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.38.0-2025.10.43-1.0