siRNA Details
| Gene Symbol: 4930506M07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114312.1 | NP_001107784.1 | 3 | 695 | ||
| NM_175172.4 | NP_780381.1 | 3 | 695 | |||
| GenBank mRNA | AK082304.1 | - | 3 | 696 | ||
| AK220405.1 | - | 3 | 725 | |||
| BC030338.1 | - | 3 | 267 | 2599101 | ||
| EF055486.1 | - | 3 | 138 | |||

4930506M07Rik
Genomic Map